Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-CD4, Clone: RPA-T4, PE,
Mouse Monoclonal Antibody
$101.07 - $747.21
Specifications
Antigen | CD4 |
---|---|
Clone | RPA-T4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1718
|
MilliporeSigma™
MABF1718 |
25 Tests |
Each for $101.07
|
|
|||||
MABF1719
|
MilliporeSigma™
MABF1719 |
100 Tests |
Each for $205.34
|
|
|||||
MABF1720
|
MilliporeSigma™
MABF1720 |
500 Tests |
Each for $747.21
|
|
|||||
Description
Specifically detects CD4 clone: RPA-T4 in Human samples, and it is validated for Flow Cytometry
The RPA-T4 antibody reacts with human CD4, a 59kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4, and co-receptors CCR5 and CXCR4, may also be utilized by HIV-1 to enter T cells. Human CD4 is typically expressed on thymocytes, some mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.The RPA-T4 antibody is widely used as a phenotypic marker for human CD4 expression, and is cross-reactive with Chimpanzee CD4. This antibody recognizes a different epitope, and thus does not block binding of, the alternative Anti-Human CD4 antibody clone OKT4 (Reinherz EL, et al. 1979. Proc. Natl. Acad. Sci. 76:4061-4065).Specifications
CD4 | |
Flow Cytometry | |
PE | |
Mouse | |
Inflammation and Immunology | |
Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
CD4 | |
Primary | |
Stable for one year at 2°C to 8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°C to 8°C and protected from prolonged exposure to light. Do not freeze. |
RPA-T4 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P01730 | |
Immunogen information for this antibody is proprietary information. | |
Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title