Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-CD8, Clone: SK1, PE,
Mouse Monoclonal Antibody
$316.00 - $3263.00
Specifications
| Antigen | CD8 |
|---|---|
| Clone | SK1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF1676
|
MilliporeSigma™
MABF1676 |
25 Tests |
Each for $316.00
|
|
|||||
|
MABF1677
|
MilliporeSigma™
MABF1677 |
100 Tests |
Each for $727.00
|
|
|||||
|
MABF1678
|
MilliporeSigma™
MABF1678 |
500 Tests |
Each for $3,263.00
|
|
|||||
Description
Specifically detects CD8 clone: SK1 in Human samples, and it is validated for Flow Cytometry
The SK1 antibody is specific for the 32-34kDa alpha chain of human CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor for antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The SK1 antibody is widely used as a phenotypic marker for CD8 on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells. It is cross-reactive with CD8 in several non-human species, including Baboon, Chimpanzee, Cynomolgus and Rhesus. If used together with an alternative Anti-Human CD8a clone, RPA-T8, the SK1 antibody will not block binding of RPA-T8 to CD8a.Specifications
| CD8 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Inflammation and Immunology | |
| Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
| CD8A; MAL | |
| Primary | |
| Stable for one year at 2°C to 8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°C to 8°C and protected from prolonged exposure to light. Do not freeze. |
| SK1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P01732 | |
| Immunogen information for this antibody is proprietary information. | |
| Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title