Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Cd8a, Clone: C8/1035, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2453270.1MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for ELISA.Specifications
CD8 alpha | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
CD8, CD8 antigen, alpha polypeptide (p32), CD8a antigen, CD8a molecule, Leu2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, RPA-T8, RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone, RPA-T8 Flow, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 | |
Mouse | |
32 kDa | |
0.1 mg | |
Innate Immunity | |
925 | |
Human | |
Purified |
Flow Cytometry, SDS-Page, Immunofluorescence | |
C8/1035 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD8A | |
Recombinant full-length human CD8a protein | |
Protein A or G purified | |
RUO | |
Primary | |
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas are CD4+/CD8-. Anaplastic large cell lymphoma is usually CD4+ and CD8-, and in T-lymphoblastic lymphoma/leukemia, CD4 and CD8 are often co-expressed. CD8 is also found in littoral cell angioma of the spleen. | |
Store at 4C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction