Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C4b/d Antibody (C4D203), Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | Complement 4d |
|---|---|
| Clone | C4D203 |
| Concentration | 0.2 mg/mL |
| Dilution | SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | SDS-Page, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24479402
![]() |
Novus Biologicals
NBP2447940.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in ELISA, Immunofluorescence
Complement C4b/d Monoclonal specifically detects Complement C4b/d in Human samples. It is validated for Immunofluorescence.Specifications
| Complement 4d | |
| 0.2 mg/mL | |
| SDS-Page, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| C4d | |
| Recombinant human Complement 4d protein | |
| Primary | |
| Store at 4C. | |
| 192 kDa |
| C4D203 | |
| SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| C4A | |
| IgG1 | |
| Protein A or G purified | |
| This MAb is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound C4d.C4d is a degradation product of the activated complement factor C4b. Complement 4b is typically activated by binding of Abs to specific target molecules. Following activation and degradation of the C4 molecule, thio-ester groups are exposed, which allow transient, covalent binding of the degradation product Complement 4d to endothelial cell surfaces and extracellular matrix components of vascular basement membranes near the sites of C4 activation. The presence of C4d in peritubular capillaries is a key indicator for acute humoral (i.e. antibody-mediated) rejection of kidney, heart, pancreas and lung allografts. As an established marker of antibody-mediated acute renal allograft rejection and its proclivity for endothelium, this component can be detected in peritubular capillaries in chronic renal allograft rejection as well as hyperacute rejection, acute vascular rejection, acute cellu |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title