Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Complement C5, Clone: 561, Novus Biologicals™
SDP

Catalog No. NBP16014705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16014705 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP16014705 Supplier Novus Biologicals Supplier No. NBP1601470.05MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Complement C5 Monoclonal specifically detects Complement C5 in Human samples. It is validated for ELISA, Functional, Immunoassay.

Specifications

Antigen Complement C5
Applications ELISA, Functional Assay, Immunoassay
Classification Monoclonal
Clone 561
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution ELISA detection, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS and 0.1% BSA with No Preservative
Gene Accession No. P01031
Gene Alias anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822
Gene Symbols C5
Host Species Mouse
Immunogen Human C5/C5a
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity Monoclonal antibody 561 recognizes the N-terminus of C5. The antibody reacts both with intact C5 (190 kDa) as with C5a (115 kDa).
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.