Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CREB (Ascites Free), Clone: LB9,

Catalog No. MAB5432C Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5432C 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5432C Supplier MilliporeSigma™ Supplier No. MAB5432C
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CREB (Ascites Free) clone: LB9 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Cyclic AMP-responsive element-binding protein 1 (UniProt P16220; also known as cAMP-responsive element-binding protein 1, CREB-1) is encoded by the CREB1 gene (Gene ID 1385) in human. CREB is a transcription factor that binds the cAMP response elements (CRE) and regulates the transcription of the downstream genes. CREB activity is regulation via phosphorylation on by various kinases, including PKA and Ca2+/calmodulin-dependent protein kinases. Genes known to be regulated by CREB include c-fos, BDNF, tyrosine hydroxylase, neuropeptides (such as somatostatin, enkephalin, VGF, and corticotropin-releasing hormone), as well as the circadian clock genes PER1 and PER2. CREB is critical for memory formation across a wide range of species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB (Ascites Free)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LB9
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P16220
Gene Symbols CREB1
Host Species Mouse
Immunogen Recombinant CREB-A protein.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_604391
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.