Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EEA1 Antibody (6D4.1D4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $438.00
Specifications
| Antigen | EEA1 |
|---|---|
| Clone | 6D4.1D4 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33212210), Immunohistochemistry-Paraffin 5-10 ug/ml, Flow (Intracellular) 2.5 ug/million cells |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP236568SS
![]() |
Novus Biologicals
NBP236568SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP236568
![]() |
Novus Biologicals
NBP236568 |
0.1 mg |
Each for $438.00
|
|
|||||
Description
EEA1 Monoclonal specifically detects EEA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
| EEA1 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Endosome Markers, Membrane Trafficking and Chaperones | |
| PBS with 0.02% Sodium Azide | |
| early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2 | |
| EEA1 | |
| IgG1 κ | |
| Protein G purified |
| 6D4.1D4 | |
| Western Blot, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33212210), Immunohistochemistry-Paraffin 5-10 ug/ml, Flow (Intracellular) 2.5 ug/million cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Virus | |
| Q15075 | |
| 8411 | |
| Partial recombinant human EEA1 protein (between amino acids 50-300). [UniProt Q15075] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title