Learn More
Description
Specifications
Specifications
| Antigen | EGFR |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | H9B4 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-1.0ug/ml, ELISA 1-5ug/ml for coating, Immunoprecipitation 1-2ug/500ug protein lysate |
| Formulation | 10mM PBS - BSA free with No Preservative |
| Gene Accession No. | P00533 |
| Gene Alias | avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
