Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Ferroportin/SLC40A1, Clone: 8G10NB, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP2453560.025MG
Description
Ensure accurate, reproducible results in Western Blot, Flow (Cell Surface), Immunohistochemistry, Immunohistochemistry (Paraffin)
Ferroportin/SLC40A1 Monoclonal specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Ferroportin/SLC40A1 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 2-4 ug/ml, Flow Cytometry, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface) 25 ug/ml, Flow (Intracellular) | |
Q9NP59 | |
SLC40A1 | |
Partial recombinant human Ferroportin protein (between amino acids 150-350) [UniProt Q9NP59] | |
0.025 mg | |
Cancer, Signal Transduction | |
30061 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
8G10NB | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4ferroportin-1, Iron-regulated transporter 1, member 3, MST079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction