Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Ferroportin/SLC40A1, Clone: 8G10NB, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$182.00 - $401.00
Specifications
Antigen | Ferroportin/SLC40A1 |
---|---|
Clone | 8G10NB |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2-4 ug/ml, Flow Cytometry, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface) 25 ug/ml, Flow (Intracellular) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24535602
![]() |
Novus Biologicals
NBP2453560.025MG |
0.025 mg |
Each for $182.00
|
|
|||||
NBP24535601
![]() |
Novus Biologicals
NBP2453560.1MG |
0.1 mg |
Each for $401.00
|
|
|||||
Description
Ensure accurate, reproducible results in Western Blot, Flow (Cell Surface), Immunohistochemistry, Immunohistochemistry (Paraffin)
Ferroportin/SLC40A1 Monoclonal specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Ferroportin/SLC40A1 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4ferroportin-1, Iron-regulated transporter 1, member 3, MST079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 | |
SLC40A1 | |
IgG2b κ | |
Protein G purified |
8G10NB | |
Western Blot 2-4 ug/ml, Flow Cytometry, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface) 25 ug/ml, Flow (Intracellular) | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q9NP59 | |
30061 | |
Partial recombinant human Ferroportin protein (between amino acids 150-350) [UniProt Q9NP59] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title