Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Glycophorin A, Clone: A63-B/C2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450210.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Erythrocyte Agglutination, Immunofluorescence
Glycophorin A Monoclonal antibody specifically detects Glycophorin A in Human, Bovine samples. It is validated for Flow Cytometry, Immunofluorescence, Erythrocyte Agglutination.Specifications
Glycophorin A | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml, Erythrocyte Agglutination | |
P02724 | |
GYPA | |
Human erythrocytes treated with neuraminidase | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a sialoglycoprotein of 39kDa, identified as glycophorin A (GPA). It is present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorin acts as the receptor for Sandei virus and parvovirus. Glycophorins A (GPA) and B (GPB), which are single, trans-membrane sialoglycoproteins. GPA is the carrier of blood group M and N specificities, while GPB accounts for S and U specificities. GPA and GPB provide the cells with a large mucin like surface and it has been suggested this provides a barrier to cell fusion, so minimizing aggregation between red blood cells in the circulation. | |
Store at 4C. | |
IgM κ |
Flow Cytometry, Immunofluorescence, Agglutination | |
A63-B/C2 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
Mouse | |
39 kDa | |
0.2 mg | |
Cancer, Signal Transduction | |
2993 | |
Human, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction