Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Glycophorin A, Clone: A63-B/C2, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | Glycophorin A |
---|---|
Clone | A63-B/C2 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml, Erythrocyte Agglutination |
Applications | Flow Cytometry, Immunofluorescence, Agglutination |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24502102
![]() |
Novus Biologicals
NBP2450210.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Erythrocyte Agglutination, Immunofluorescence
Glycophorin A Monoclonal antibody specifically detects Glycophorin A in Human, Bovine samples. It is validated for Flow Cytometry, Immunofluorescence, Erythrocyte Agglutination.Specifications
Glycophorin A | |
0.2 mg/mL | |
Flow Cytometry, Immunofluorescence, Agglutination | |
Unconjugated | |
Mouse | |
Cancer, Signal Transduction | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
GYPA | |
IgM κ | |
Protein A or G purified | |
Recognizes a sialoglycoprotein of 39kDa, identified as glycophorin A (GPA). It is present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorin acts as the receptor for Sandei virus and parvovirus. Glycophorins A (GPA) and B (GPB), which are single, trans-membrane sialoglycoproteins. GPA is the carrier of blood group M and N specificities, while GPB accounts for S and U specificities. GPA and GPB provide the cells with a large mucin like surface and it has been suggested this provides a barrier to cell fusion, so minimizing aggregation between red blood cells in the circulation. |
A63-B/C2 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml, Erythrocyte Agglutination | |
Monoclonal | |
Purified | |
RUO | |
Human, Bovine | |
P02724 | |
2993 | |
Human erythrocytes treated with neuraminidase | |
Primary | |
Store at 4C. | |
39 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title