Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Glycophorin A, Clone: SPM183, Novus Biologicals™
SDP

Catalog No. NBP24502002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24502002 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24502002 Supplier Novus Biologicals Supplier No. NBP2450200.2MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Flow Cytometry, Erythrocyte Agglutination, Immunofluorescence

Glycophorin A Monoclonal antibody specifically detects Glycophorin A in Human, Bovine samples. It is validated for Flow Cytometry, Immunofluorescence, Erythrocyte Agglutination.

Specifications

Antigen Glycophorin A
Applications Flow Cytometry, Immunofluorescence, Agglutination
Classification Monoclonal
Clone SPM183
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml, Erythrocyte Agglutination
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P02724
Gene Alias CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR
Gene Symbols GYPA
Host Species Mouse
Immunogen Human erythrocytes treated with neuraminidase
Molecular Weight of Antigen 39 kDa
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2993
Test Specificity Recognizes a sialoglycoprotein of 39kDa, identified as glycophorin A. It reacts with a peptide epitope on the extracellular domain of human glycophorin. Glycophorin A is present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorins acts as the receptor for Sandei virus and parvovirus.
Target Species Human, Bovine
Content And Storage Store at 4C.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.