Learn More
HLA-DR Mouse anti-Human, APC-R700, Clone: G46-6, BD
Mouse Monoclonal Antibody
$186.20 - $366.00
Specifications
| Antigen | HLA-DR |
|---|---|
| Clone | G46-6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-R700 |
Description
The G46-6 monoclonal antibody specifically binds to HLA-DR, a major histocompatibility complex (MHC) class II antigen. HLA-DR antigens are encoded by genes within the Human Leukocyte Antigen (HLA) Complex located on chromosome 6. HLA-DR is a transmembrane heterodimeric glycoprotein composed of an α chain (36 kDa) and a β subunit (27 kDa) expressed primarily on antigen presenting cells: B cells, dendritic cells, monocytes, macrophages, and thymic epithelial cells. HLA-DR is also expressed on activated T cells. This molecule plays a major role in mediating cellular interactions during antigen presentation to CD4-positive T cells.
Specifications
| HLA-DR | |
| Flow Cytometry | |
| APC-R700 | |
| RUO | |
| Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| G46-6 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG2a κ | |
| Affinity Purified | |
| MHC class II antigen; HLA class II histocompatibility antigen |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.