Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgM Fc Secondary Antibody (CH2), PerCP, Novus Biologicals™
SDP

Catalog No. NB500468PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500468PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500468PCP Supplier Novus Biologicals Supplier No. NB500468PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgM Fc Monoclonal antibody specifically detects IgM Fc in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen IgM Fc
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CH2
Conjugate PerCP
Dilution Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500
Host Species Mouse
Immunogen Purified human IgM.
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The antibody CH2 reacts with Fc fragment of human IgM.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.