Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-IL-3 R alpha/CD123, Clone: 6H6, Cy5.5, PerCP, Novus Biologicals™
SDP

Catalog No. NBP20046825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP20046825 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP20046825 Supplier Novus Biologicals Supplier No. NBP20046825 TESTS

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IL-3 R alpha/CD123 Monoclonal antibody specifically detects IL-3 R alpha/CD123 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-3 R alpha/CD123
Applications Flow Cytometry
Classification Monoclonal
Clone 6H6
Conjugate PerCP-Cyanine5.5
Dilution Flow Cytometry 1:10-1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Gene Alias CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Gene Symbols IL3RA
Host Species Mouse
Immunogen Tissue/ cell preparation. IL3R transfected COS cells.
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3563
Test Specificity IL3RA (6H6)
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.