Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Integrin alpha M/CD11b, Clone: C11b/660, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2345730.2 MG
Description
CD11b Monoclonal antibody specifically detects CD11b in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready.Specifications
CD11b | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Frozen 0.5-1ug/ml, Functional, CyTOF-ready | |
P11215 | |
ITGAM | |
Recombinant human CD11b protein | |
Protein A purified | |
RUO | |
Primary | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
C11b/660 | |
Unconjugated | |
10mM PBS - BSA free with No Preservative | |
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, CD11Bintegrin, alpha M (complement component receptor 3, alpha; also known as CD11b(p170), macrophage antigen alpha polypeptide), Cell surface glycoprotein MAC-1 subunit alpha, CR-3 alpha chain, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
Mouse | |
127 kDa | |
0.2 mg | |
Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
3684 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction