Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Integrin beta 3/CD61, Clone: 25E11., Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | Integrin beta 3/CD61 |
---|---|
Clone | 25E11. |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24496402
![]() |
Novus Biologicals
NBP2449640.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
Integrin beta 3/CD61 | |
0.2 mg/mL | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Cytoskeleton Markers, Signal Transduction | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
ITGB3 | |
IgM κ | |
Protein A or G purified | |
Reacts with human integrin beta3 (GPIIIa, vitronectin receptor beta chain). The protein detectable is a complex of CD41 and CD61.The apparent molecular weight of the GPIIIa by SDS-PAGE is 105kDa reduced and 90kDa unreduced. Ligands are fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin. Residues 237-248 of GPIIIa or CD61 are critical in adhesive protein binding |
25E11. | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P05106 | |
3690 | |
Normal blood mononuclear cells activated by mixed lymphocyte reaction | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title