Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LNPEP, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104850 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-850 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-850 Supplier Abnova Corporation Supplier No. H00004012A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant LNPEP.

This gene encodes a zinc-dependent aminopeptidase that cleaves vasopressin, oxytocin, lys-bradykinin, met-enkephalin, dynorphin A and other peptide hormones. The protein can be secreted in maternal serum, reside in intracellular vesicles with the insulin-responsive glucose transporter GLUT4, or form a type II integral membrane glycoprotein. The protein catalyzes the final step in the conversion of angiotensinogen to angiotensin IV (AT4) and is also a receptor for AT4. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: FIIRTVGRHFPGHLLAWDFVKENWNKLVQKFPLGSYTIQNIVAGSTYLFSTKTHLSEVQAFFENQSEATFRLRCVQEALEVIQLNIQWMEKNLKSLTWWL

Specifications

Antigen LNPEP
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant LNPEP.
Formulation 50% glycerol
Gene LNPEP
Gene Accession No. NM_005575
Gene Alias CAP/IRAP/P-LAP/PLAP
Gene Symbols LNPEP
Host Species Mouse
Immunogen LNPEP (NP_005566, 926 a.a. ∽ 1025 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4012
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.