Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Melanoma Marker (MART-1 + gp100), Clone: DT101+ BC199 + HMB45, Novus Biologicals™
SDP

Catalog No. NBP23460802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23460802 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23460802 Supplier Novus Biologicals Supplier No. NBP2346080.2 MG

Mouse Monoclonal Antibody

Melanoma Marker (MART-1 + gp100) Monoclonal antibody specifically detects Melanoma Marker (MART-1 + gp100) in Human samples. It is validated for Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Melanoma Marker (MART-1 + gp100)
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone DT101+ BC199 + HMB45
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml
Formulation 10mM PBS - BSA free with No Preservative
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein (DT101& BC199); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1, IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.