Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Melanoma Marker (MART-1 + Tyrosinase + gp100), Clone: A103 + T311 + HMB45, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | Melanoma Marker (MART-1 + Tyrosinase + gp100) |
---|---|
Clone | A103 + T311 + HMB45 |
Concentration | 1.0 mg/mL |
Dilution | Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23454702
![]() |
Novus Biologicals
NBP2345470.2 MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Melanoma Marker (MART-1 + Tyrosinase + gp100) Monoclonal antibody specifically detects Melanoma Marker (MART-1 + Tyrosinase + gp100) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Melanoma Marker (MART-1 + Tyrosinase + gp100) | |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human | |
Q16655 | |
Recombinant hMART-1 protein (A103); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45) | |
Primary | |
Protein A purified | |
This antibody cocktail recognizes three melanoma-specific proteins, which include MART-1, Tyrosinase and gp100. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. The function of gp100 is not known but it is reported to be a useful marker for melanocytes and melanomas. This cocktail of three markers is designed for extremely sensitive labeling of formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation. |
A103 + T311 + HMB45 | |
Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS - BSA free with No Preservative | |
2315 | |
IgG1, IgG2a κ | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title