Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-NRAS, Clone: 2G9, Novus Biologicals™
SDP

Catalog No. NBP17404410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17404410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17404410 Supplier Novus Biologicals Supplier No. NBP174044100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NRAS Monoclonal specifically detects NRAS in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NRAS
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2G9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100
Formulation PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Gene Accession No. NP_002515
Gene Alias ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog
Gene Symbols NRAS
Host Species Mouse
Immunogen Recombinant human NRAS (1-186aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4893
Test Specificity Anti-human RAS mAb, clone 2G9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NRAS protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.