Learn More
Mouse anti-Nucleoli Marker, Clone: NM95, Azide Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Nucleoli Marker |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | NM95 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml |
| Formulation | 10mM PBS - BSA free with No Preservative |
| Host Species | Mouse |
| Immunogen | Nuclei of myeloid leukemia biopsy cells |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.