Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Osteopontin/OPN, Clone: 53, Novus Biologicals™
SDP

Catalog No. NBP11918405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP11918405 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP11918405 Supplier Novus Biologicals Supplier No. NBP1191840.05MG

Mouse Monoclonal Antibody

Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Osteopontin/OPN
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 53
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with 0.1% Sodium Azide
Gene Accession No. p10451
Gene Alias BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin
Gene Symbols SPP1
Host Species Mouse
Immunogen Recombinant human Osteopontin
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Extracellular Matrix, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 6696
Test Specificity Osteopontin (53)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.