Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-PGP9.5 / UCHL-1, Clone: 13C4, Novus Biologicals™
SDP

Catalog No. NBP22942002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22942002 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22942002 Supplier Novus Biologicals Supplier No. NBP2294200.2MG

Specifications

Antigen UCH-L1/PGP9.5
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 13C4
Concentration 0.2mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml, Simple Western 10 ug/ml, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P09936
Gene Alias EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
Gene Symbols UCHL1
Host Species Mouse
Immunogen PGP9.5 protein from human brain was used as the immunogen for the PGP9.5 antibody.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 7345
Test Specificity This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig, Rabbit, Sheep, Zebrafish
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.