Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG Secondary Antibody (MR12), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP25026105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25026105 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP25026105 Supplier Novus Biologicals Supplier No. NBP250261AF405

Mouse Monoclonal Antibody

IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Rabbit IgG
Applications Immunohistochemistry
Classification Monoclonal
Clone MR12
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Immunoglobulin purified from rabbit serum
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Human, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.