Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Renal Cell Carcinoma (gp200), Clone: 66.4.C2 (PN-15), Azide Free, Novus Biologicals™
SDP

Catalog No. NBP23315202 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23315202 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23315202 Supplier Novus Biologicals Supplier No. NBP2331520.2MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen Renal Cell Carcinoma (gp200)
Applications ELISA
Classification Monoclonal
Clone 66.4.C2 (PN-15)
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1-5ug/ml for coating
Formulation 10mM PBS - BSA free with No Preservative
Gene Alias CD205LY-75, CLEC13BLy-75, C-type lectin domain family 13 member B, DEC-205CD205 antigen, GP200-MR6, lymphocyte antigen 75
Gene Symbols LY75
Host Species Mouse
Immunogen Fresh, normal human renal cortical tissue homogenate was used as immunogen to generate the RCC antibody.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primary or Secondary Primary
Gene ID (Entrez) 4065
Test Specificity Recognizes a glycoprotein of ∼200kDa, identified as carbonic anhydrase IX (CAIX/gp200). Its epitope resides in the carbohydrate domain of gp200. It shows no significant cross-reactivity with other carbohydrate determinants, such as the Lewis blood group antigens, epithelial membrane antigen, HMFG, and AB blood group antigens. In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas. This MAb may be useful in the investigations of carcinomas of proximal nephrogenic differentiation especially those showing tubular differentiation.
Target Species Human, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.