Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sec8 Antibody (14G1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP1974920.025MG
Description
Sec8 Monoclonal specifically detects Sec8 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Sec8 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| EXOC4 | |
| Recombinant rat rSec8. | |
| 0.025 mg | |
| Primary | |
| Bovine, Canine, Fish, Hamster, Primate, Guinea Pig, Rabbit, Sheep, Mouse, Human, Rat, Chicken | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 14G1 | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 12.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
| exocyst complex component 4, KIAA1699SEC8L1, MGC27170, REC8, SEC8Exocyst complex component Sec8, SEC8-like 1, SEC8-like 1 (S. cerevisiae), Sec8p | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 60412 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction