Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-SIRP alpha/CD172a, Clone: P3C4, Novus Biologicals™
SDP

Catalog No. NBP15455305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15455305 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP15455305 Supplier Novus Biologicals Supplier No. NBP1545530.05MG

Specifications

Antigen SIRP alpha/CD172a
Applications Flow Cytometry
Classification Monoclonal
Clone P3C4
Concentration 1mg/mL
Conjugate Unconjugated
Formulation PBS containing 50% glycerol, pH 7.2.
Host Species Mouse
Immunogen This hybridoma was established by fusion of mouse myeloma cell SP2/0 with Balb/c mouse splenocyte immunized with recombinant GST fusion containing only the N-terminal Immunoglobulin-like domain of SIRP alpha1.
Purification Method Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140885
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.