Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA4, Mouse, Clone: BRG-01, Abnova™

Catalog No. 89112878 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-878 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-878 Supplier Abnova Corporation Supplier No. MAB6460
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SMARCA4.

The protein encoded by this gene is a member of the SWI/SNF family of proteins and is similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. In addition, this protein can bind BRCA1, as well as regulate the expression of the tumorigenic protein CD44. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen SMARCA4
Applications Western Blot
Classification Monoclonal
Clone BRG-01
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SMARCA4.
Dilution Western Blot (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene SMARCA4
Gene Alias BAF190/BRG1/FLJ39786/SNF2/SNF2-BETA/SNF2L4/SNF2LB/SWI2/hSNF2b
Gene Symbols SMARCA4
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to C-terminus 242 amino acids of human SMARCA4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6597
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.