Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYN2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105543 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
89-105-543 50 μL
1 options

Catalog No. 89-105-543

Supplier: Abnova Corporation H00006854A01

Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SYN2.

This gene is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. This member of the synapsin family encodes a neuron-specific phosphoprotein that selectively binds to small synaptic vesicles in the presynaptic nerve terminal. The TIMP4 gene is located within an intron of this gene and is transcribed in the opposite direction. Mutations in this gene may be associated with abnormal presynaptic function and schizophrenia. Alternative splicing of this gene results in two transcripts. [provided by RefSeq

Sequence: AMSDRYKLWVDTCSEMFGGLDICAVKAVHGKDGKDYIFEVMDCSMPLIGEHQVEDRQLITELVISKMNQLLSRTPALSPQRPLTTQQPQSGTLKDPDSSKT

Specifications

Antigen SYN2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SYN2.
Formulation 50% glycerol
Gene SYN2
Gene Accession No. NM_133625
Gene Alias SYNII/SYNIIa/SYNIIb
Gene Symbols SYN2
Host Species Mouse
Immunogen SYN2 (NP_598328, 348 a.a. ∽ 448 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6854
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.