Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Syntaxin 6, Clone: 3D10, Novus Biologicals™
SDP

Catalog No. NBP15447805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15447805 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP15447805 Supplier Novus Biologicals Supplier No. NBP1544780.05MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Syntaxin 6 Monoclonal specifically detects Syntaxin 6 in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Syntaxin 6
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 3D10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunoprecipitation 2 ug/300 uL of cell extract
Formulation PBS (pH 7.2) and 50% Glycerol with No Preservative
Gene Accession No. Q63635
Gene Alias syntaxin 6, syntaxin-6
Gene Symbols STX6
Host Species Mouse
Immunogen A hybridoma was established by fusion of mouse myeloma cell NS-1 with Balb/c mouse splenocyte immunized with the recombinant rat syntaxin-6.
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 10228
Test Specificity It is reported that this monoclonal antibody (3D10) binds to the amino-terminal 25 amino acid of rat syntaxin-6.
Reconstitution Protein A purified
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.