Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin C Antibody (T2H5), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$280.00 - $524.00

Specifications

Antigen Tenascin C
Clone T2H5
Concentration 0.2mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24458200
SDP
View Documents
Novus Biologicals
NBP2445820.02MG
0.02 mg
Each for $280.00
Only null left
Add to Cart
 
NBP24458201
SDP
View Documents
Novus Biologicals
NBP2445820.1MG
0.1 mg
Each for $524.00
Only null left
Add to Cart
 
NBP24458202
SDP
View Documents
Novus Biologicals
NBP2445820.2MG
0.2 mg N/A N/A N/A
Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Tenascin C Monoclonal specifically detects Tenascin C in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Tenascin C
0.2mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Mouse
Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer
1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide
150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP
TNC
IgG1 κ
Protein A or G purified
In Western blotting, it reacts with two bands of ∼MW of 210kDa and 300kDa, identified as two isoforms of Tenascin C. Specificity of this MAb is validated by sequential immunoprecipitation with a PAb against Tenascin C. Tenascin C is a multifunctional, disulfide-linked hexameric extracellular matrix glycoprotein expressed in association with mesenchymal epithelial interactions during development and in the neo-vasculature and stroma of undifferentiated tumors. In adults, it is restricted to certain epithelial-stromal interfaces and increases markedly in hyper-proliferative diseases and in stroma of many neoplasms, including gliomas, breast, squamous and lung carcinomas.
T2H5
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Monoclonal
Purified
RUO
Human, Rat (Negative)
P24821
3371
Human breast carcinoma
Primary
Store at 4C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.