Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin C Antibody (T2H5), Novus Biologicals™
SDP

Catalog No. NBP24458201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24458201 0.1 mg
NBP24458200 0.02 mg
NBP24458202 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24458201 Supplier Novus Biologicals Supplier No. NBP2445820.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Tenascin C Monoclonal specifically detects Tenascin C in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Tenascin C
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone T2H5
Concentration 0.2mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P24821
Gene Alias 150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP
Gene Symbols TNC
Host Species Mouse
Immunogen Human breast carcinoma
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3371
Test Specificity In Western blotting, it reacts with two bands of ∼MW of 210kDa and 300kDa, identified as two isoforms of Tenascin C. Specificity of this MAb is validated by sequential immunoprecipitation with a PAb against Tenascin C. Tenascin C is a multifunctional, disulfide-linked hexameric extracellular matrix glycoprotein expressed in association with mesenchymal epithelial interactions during development and in the neo-vasculature and stroma of undifferentiated tumors. In adults, it is restricted to certain epithelial-stromal interfaces and increases markedly in hyper-proliferative diseases and in stroma of many neoplasms, including gliomas, breast, squamous and lung carcinomas.
Target Species Human, Rat (Negative)
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.