Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 1, 2, 3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5380133
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1835SP 25 μg
MAB1835100 100 μg
MAB1835500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1835SP Supplier R&D Systems Supplier No. MAB1835SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 143 publications

TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-Species samples. It is validated for Western Blot, Binding Inhibition, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TGF beta 1/2/3
Applications Western Blot, In vitro Assay, Neutralization, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1D11
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Binding Inhibition, Neutralization 0.25-1.25 μg/mL, ELISA Capture (Matched Antibody Pair) 2-8 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1
Gene Symbols TGFB1
Host Species Mouse
Immunogen Bovine bone-derived TGF-beta 1 and TGF-beta 2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Test Specificity Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3. No cross-reactivity was observed with recombinant LAP in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.