Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Thymidylate Synthase, Clone: TS106 + TMS715, Azide Free, Novus Biologicals™
SDP

Catalog No. NBP23450302 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23450302 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23450302 Supplier Novus Biologicals Supplier No. NBP2345030.2 MG

Mouse Monoclonal Antibody

Thymidylate Synthase Monoclonal antibody specifically detects Thymidylate Synthase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen Thymidylate Synthase
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TS106 + TMS715
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.25-0.5ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml, CyTOF-ready
Formulation 10mM PBS - BSA free with No Preservative
Gene Accession No. P04818
Gene Alias EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422
Gene Symbols TYMS
Host Species Mouse
Immunogen Recombinant human thymidylate synthase
Molecular Weight of Antigen 36 kDa
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7298
Test Specificity It recognizes a protein of 36kDa, identified as Thymidylate Synthase (TS) (EC 2.1.1.45). TS converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is also a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.