Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-TLR4, Clone: TLR4/230, Novus Biologicals™

Mouse Monoclonal Antibody
$244.00 - $488.00
Specifications
Antigen | TLR4 |
---|---|
Clone | TLR4/230 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24517000
![]() |
Novus Biologicals
NBP2451700.02MG |
0.02 mg |
Each for $244.00
|
|
|||||
NBP24517001
![]() |
Novus Biologicals
NBP2451700.1MG |
0.1 mg |
Each for $488.00
|
|
|||||
NBP24517002
![]() |
Novus Biologicals
NBP2451700.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
TLR4 Monoclonal specifically detects TLR4 in Human, Rat, Porcine, Canine, Guinea Pig, Monkey samples. It is validated for Functional.Specifications
TLR4 | |
0.2 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
TLR4 | |
IgG2a κ | |
Protein A or G purified | |
This MAb reacts with human Toll-like receptor 2 (TLR4). It is a member of the Toll-like receptor (TLR) family, which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This receptor has been implicated in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria. Mutations in this gene have been associated with differences in LPS responsiveness. Multiple transcript variants encoding different isoforms have been found for this gene. |
TLR4/230 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Pig, Canine, Guinea Pig, Monkey | |
O00206 | |
7099 | |
Recombinant human TLR4 protein | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title