Learn More
Mouse anti-VEGF R1/Flt-1, Clone: FLT1/659, Azide Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | VEGFR1/Flt-1 |
| Applications | ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | FLT1/659 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA 1-5ug/ml for coating, Functional |
| Formulation | 10mM PBS - BSA free with No Preservative |
| Gene Accession No. | P17948 |
| Gene Alias | EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.