Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Artemin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
$208.00 - $1973.00
Specifications
Antigen | Artemin |
---|---|
Clone | 185234 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10851SP
![]() |
R&D Systems
MAB10851SP |
25 μg |
Each for $208.00
|
|
|||||
MAB10851100
![]() |
R&D Systems
MAB10851100 |
100 μg |
Each for $558.00
|
|
|||||
MAB10851500
![]() |
R&D Systems
MAB10851500 |
500 μg |
Each for $1,973.00
|
|
|||||
Description
Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Artemin | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin | |
ARTN | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
185234 | |
Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
Q9Z0L2.1 | |
9048 | |
E. coli-derived recombinant mouse Artemin Ala112-Gly224 Accession # Q9Z0L2.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Artemin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) AgRP, rmCripto, recombinant human (rh) Cripto-1, recombinant drosophila Dpp, recombinant rat GDNF, rhLAP, rhLatent TGF-beta 1, rhLefty A, rmLefty-1, rmMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant Amphibian TGF-beta 5 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title