Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ASCL1/Mash1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7061200
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2567SP 25 μg
AF2567 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2567SP Supplier R&D Systems Supplier No. AF2567SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

ASCL1/Mash1 Polyclonal specifically detects ASCL1/Mash1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ASCL1/Mash1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q53WV9
Gene Alias achaete-scute complex (Drosophila) homolog-like 1, achaete-scute complex homolog 1 (Drosophila), achaete-scute complex-like 1 (Drosophila), achaete-scute homolog 1, ASCL1, ASH1, ASH-1, ASH1MASH1, BHLHA46, bHLHa46achaete-scute complex-like 1, Class A basic helix-loop-helix protein 46, HASH1, HASH1achaete scute protein, Mash1
Gene Symbols ASCL1
Host Species Goat
Immunogen E. coli-derived recombinant mouse ASCL1/Mash1 His74-Asp173 (Gln160Glu) Accession # Q53WV9
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 429
Test Specificity Detects mouse ASCL1/Mash1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.