Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL11/Eotaxin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
$200.50 - $2070.00
Specifications
Antigen | CCL11/Eotaxin |
---|---|
Clone | 42285 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-30 ug/mL |
Applications | Western Blot, Immunohistochemistry, Neutralization |
Classification | Monoclonal |
Description
CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
CCL11/Eotaxin | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-30 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
P48298 | |
6356 | |
E. coli-derived recombinant mouse CCL11/Eotaxin His24-Pro97 Accession # P48298 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CCL11/Eotaxin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant human (rh) CCL11/Eotaxin, recombinant mouse CCL7, and rhCCL21 is observed. |
42285 | |
Western Blot, Immunohistochemistry, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
CCL11 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title