Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCR10 Antibody, R&D Systems™

Rat Monoclonal Antibody
$214.00 - $514.50
Specifications
Antigen | CCR10/GPR2 |
---|---|
Clone | 248918R |
Dilution | Flow Cytometry 0.25μg/106 cells, CyTOF-ready |
Applications | CyTOF, Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2815RSP
![]() |
R&D Systems
MAB2815RSP |
25 μg |
Each for $214.00
|
|
|||||
MAB2815R100
![]() |
R&D Systems
MAB2815R100 |
100 μg |
Each for $514.50
|
|
|||||
Description
CCR10/GPR2 Monoclonal specifically detects CCR10/GPR2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CCR10/GPR2 | |
Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Mouse | |
NP_031747 | |
2826 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
248918R | |
CyTOF, Flow Cytometry | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2 | |
C6 rat glioma cell line transfected with mouse CCR10 Met1-Asn362 Accession # NP_031747 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects mouse CCR10 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title