Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD39L3/ENTPD3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4464SP
This item is not returnable.
View return policy
Description
CD39L3/ENTPD3 Polyclonal specifically detects CD39L3/ENTPD3 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
CD39L3/ENTPD3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ATPDase, CD39 antigen-like 3, CD39L3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, ENTPD3, FLJ93839, HB6, HB6NTPDase 3, NTPDase-3 | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
956 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunoprecipitation 25 ug/mL | |
AAH79871 | |
ENTPD3 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD39L3/ENTPD3 Gln44-Pro485 Accession # AAH79871 | |
25 μg | |
Primary | |
Detects mouse CD39L3/ENTPD3 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human CD39L3 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction