Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD39L3/ENTPD3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | CD39L3/ENTPD3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
CD39L3/ENTPD3 Polyclonal specifically detects CD39L3/ENTPD3 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
CD39L3/ENTPD3 | |
Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
ATPDase, CD39 antigen-like 3, CD39L3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, ENTPD3, FLJ93839, HB6, HB6NTPDase 3, NTPDase-3 | |
ENTPD3 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
AAH79871 | |
956 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD39L3/ENTPD3 Gln44-Pro485 Accession # AAH79871 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse CD39L3/ENTPD3 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human CD39L3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title