Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CX3CR1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$212.02 - $520.12
Specifications
| Antigen | CX3CR1 |
|---|---|
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB5825G025
![]() |
R&D Systems
FAB5825G025 |
25 Tests |
Each for $212.02
|
|
|||||
FAB5825G100
![]() |
R&D Systems
FAB5825G100 |
100 Tests |
Each for $520.12
|
|
|||||
Description
CX3CR1 Polyclonal specifically detects CX3CR1 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CX3CR1 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Mouse | |
| Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1 | |
| CX3CR1 | |
| IgG | |
| Affinity Purified | |
| Detects human and mouse CX3CR1 in direct ELISAs and Western blots. |
| Flow Cytometry 5 uL/10^6 cells | |
| Polyclonal | |
| Goat | |
| Q9Z0D9 | |
| 1524 | |
| E. coli-derived recombinant mouse CX3CR1 Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274 Accession # Q9Z0D9 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title