Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL15/Lungkine Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $655.00
Specifications
Antigen | CXCL15/Lungkine |
---|---|
Clone | 96708 |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
CXCL15/Lungkine Monoclonal specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
CXCL15/Lungkine | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
lungkine, Scyb15, weche | |
CXCL15 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
96708 | |
Western Blot, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q9WVL7 | |
20309 | |
E. coli-derived recombinant mouse CXCL15/Lungkine Gln26-Ala167 Accession # Q9WVL7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CXCL15/Lungkine in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) CXCL13, recombinant human (rh) CXCL13, rhCXCL5, rhCXCL8, rmMIP-2, recombinant rat CINC-2 beta , rmCX3CL1, rmCCL12, or rhCCL15 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title