Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Desert Hedgehog/Dhh N-Terminus Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Desert Hedgehog/Dhh |
---|---|
Clone | 126925 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Desert Hedgehog/Dhh Monoclonal specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot.Specifications
Desert Hedgehog/Dhh | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, DHH, HHG-3MGC35145 | |
DHH | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
126925 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q61488 | |
50846 | |
E. coli-derived recombinant mouse Desert Hedgehog/Dhh Cys23-Gly396 Accession # Q61488 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Desert Hedgehog/Dhh N-Terminus in direct ELISAs and Western blots. In Western blots, no cross-reactivity with the N-terminal peptides of recombinant human Shh or recombinant mouse (rm) Shh is observed. Also, in direct ELISAs and Western blots, no cross-reactivity with the C-terminal peptides of rmDhh, rmIhh, or rmShh is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title