Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DPPII/QPP/DPP7 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | DPPII/QPP/DPP7 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization |
Classification | Polyclonal |
Description
DPPII/QPP/DPP7 Polyclonal specifically detects DPPII/QPP/DPP7 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
DPPII/QPP/DPP7 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
carboxytripeptidase, Dipeptidyl aminopeptidase II, dipeptidyl arylamidase II, dipeptidyl peptidase 2, Dipeptidyl peptidase 7, Dipeptidyl peptidase II, dipeptidylpeptidase 7, dipeptidyl-peptidase 7, dipeptidyl-peptidase II, DPP II, DPP2, DPP7, DPPII, EC 3.4.14.2, QPP, Quiescent cell proline dipeptidase | |
DPP7 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
RUO | |
NP_114031 | |
29952 | |
Mouse myeloma cell line NS0-derived recombinant mouse DPPII/QPP/DPP7 Asp34-Arg506 Accession # NP_114031 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse DPPII/QPP/DPP7 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human DPPII is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title