Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse E-Selectin/CD62E Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5751500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5751500 500 μg
MAB5751SP 25 μg
MAB5751100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB5751500 Supplier R&D Systems Supplier No. MAB5751500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen E-Selectin/CD62E
Applications Western Blot, ELISA
Classification Monoclonal
Clone 96419
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q00690
Gene Alias CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E
Gene Symbols SELE
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse E-Selectin Trp22-Pro557 Accession # Q00690
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6401
Test Specificity Detects mouse E-Selectin in ELISAs and Western blots. Does not cross-react with recombinant human (rh) E-Selectin, rmL-Selectin, or rhP-Selectin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.