Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse EphA4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7066846
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB641SP 25 μg
MAB641 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB641SP Supplier R&D Systems Supplier No. MAB641SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

EphA4 Monoclonal specifically detects EphA4 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen EphA4
Applications Western Blot
Classification Monoclonal
Clone 107925
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q03137
Gene Alias Cek8, EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, Sek1, Tyro1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1
Gene Symbols EPHA4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphA4 Val20-Thr547 Accession # Q03137
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2043
Test Specificity Detects mouse EphA4 in direct ELISAs and Western blots. Shows less than 5% cross-reactivity with recombinant mouse (rm) EphA3 and recombinant rat (rr) EphB1 and no cross-reactivity with recombinant human EphA1, rmEphA2, -A6, -A7, -A8, -B2, -B3, -B4, -B6, or rrEphA5.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.